Tisk…

Anti-Wee1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3120-100EA 8430 CZK
BIOV3120-100
Anti-Wee1 Rabbit Polyclonal Antibody
Wee1 is a nuclear tyrosine protein kinase. The protein plays a role in suppressing eukaryotes’ entry into mitosis by inactivating cdc2 kinase by inhibiting tyrosine phosphorylation. The predicted molecular weight of the kinase is 72 kDa. The kinase possesses tyrosine kinase activity, but structurally belongs to the serine/threonine protein kinase family.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in in Immunoprecipitation: 5-10 µg/ml

Type: Primary
Antigen: Wee1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR