Tisk…

Anti-HNF4A Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3121-100EA 10350 CZK
BIOV3121-100
Anti-HNF4A Rabbit Polyclonal Antibody
HNF4A is a nuclear transcription factor which binds DNA as a homodimer. This protein controls the expression of several genes, including hepatocyte nuclear factor 1 alpha, a transcription factor which regulates the expression of several hepatic genes. This protein may play a role in development of the liver, kidney, and intestines. Mutations in the HNF4A gene have been associated with monogenic autosomal dominant non-insulin-dependent diabetes mellitus type I.

Recommended Dilutions: ELISA: 0.25 ?g/ml; Western Blot: 2.5-5.0 ?g/ml; Iimmunofluorescence: 1:10-1:50; Flow Cytometry: 1:10-1:50).

Type: Primary
Antigen: HNF4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR