Tisk…

Anti-Caspase-9 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3136-100EA 8120 CZK
BIOV3136-100
Anti-Caspase-9 Rabbit Polyclonal Antibody
Caspase family of cysteine proteases plays a key role in apoptosis. Caspase-9 is one of the most important caspases among the caspase family members. Upon induction of apoptosis, cytochrome c released from mitochondria associates with procaspase-9/Apaf-1. The complex processes procaspase-9 into a large subunit (35 kDa or 17-25 kDa) and small (10 kDa) by self-cleavage at D315. Activated caspase-9 further cleaves other caspase members including caspase-3, one of the proteases responsible for the proteolytic cleavage of many key proteins in apoptosis. In addition to self-cleavage, procaspase-9 can also be cleaved in vivo by caspase-3 at D330. The process served as a positive feedback to amplify the apoptotic signal in caspase activation pathway.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Caspase-9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR