Tisk…

Anti-Hexokinase III Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3143-100EA 9170 CZK
BIOV3143-100
Anti-Hexokinase III Rabbit Polyclonal Antibody
Protilátky
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, thus committing glucose to the glycolytic pathway. This gene encodes hexokinase 3. Similar to hexokinases 1 and 2, this allosteric enzyme is inhibited by its product glucose-6-phosphate.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100 - 1:500; IHC (1:50 - 1:100).

Type: Primary
Antigen: Hexokinase III
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR