Tisk…

Anti-PTEN Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3479-100EA 8430 CZK
BIOV3479-100
Anti-PTEN Rabbit Polyclonal Antibody
PTEN (phosphatase and tensin homologue deleted on chromosome ten) phosphatase is a tumor suppressor gene implicated in a wide variety of human cancers. PTEN encodes the 403 amino acid polypeptide originally described as a dual-specificity protein phosphatase. The main substrates of PTEN are inositol phospholipids generated by the activation of the phosphoinositide 3 kinase (PI3K;PTEN is a major negative regulator of the PI3K/Akt signaling pathway.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunoprecipitation: 10-20 µg/ml

Type: Primary
Antigen: PTEN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR