Tisk…

Anti-Presenilin 1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3481-100EA 7470 CZK
BIOV3481-100
Anti-Presenilin 1 Rabbit Polyclonal Antibody
Protilátky
Presenilin 1 and presenilin 2 are transmembrane proteins belonging to the presenilin family. Mutation of presenilin genes has been linked to early onset of Alzheimer’s disease, probably functioning throμgh its associated g-secretase activity for amyloid-β protein processing. Endogenous presenilin mainly exists in a heterodimeric complex formed from the endoproteolytically processed N-terminal (34 kDa) and C-terminal (~20 kDa) fragments.

Recommended Dilutions: Western Blot: 2-4 µg/ml

Type: Primary
Antigen: Presenilin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR