Tisk…

Anti-EG-VEGF Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3489R-100EA 6910 CZK
BIOV3489R-100
Anti-EG-VEGF Rabbit Polyclonal Antibody
Protilátky
Human EG-VEGF (Endocrine-gland-derived vascular endothelial growth factor) is a 9.6 kDa protein consisting of 86 amino acid residues. Human EG-VEGF induces proliferation, migration and fenestration in capillary endothelial cells derived from endocrine glands.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: EG-VEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR