Tisk…

Anti-FIAF Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3490-100EA 7470 CZK
BIOV3490-100
Anti-FIAF Rabbit Polyclonal Antibody
Protilátky
Adipose tissue is an energy reserve in animals. Adipose cells produce and secrete numerous physiologically important proteins that help regulate whole body metabolism, such as adiponectin leptin. Fasting causes significant changes in nutrient metabolism and many of these changes are controlled by transcription factors that regulate rate-limiting enzymes. An important transcription factor that mediates metabolic processes induced by fasting is peroxisome proliferator-activated receptor alpha (PPAR alpha;PPAR alpha has recently been reported to target a novel gene encoding the secreted protein FIAF (fasting-induced adipose factor;FIAF is expressed predominantly in adipose tissue and is strongly upregulated in white adipose tissue and the liver under fasting conditions. Initial studies sμggest that FIAF represents a novel endocrine signal helping to regulate metabolism, especially under fasting conditions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;It recognizes ~50 kDa FIAF from samples of mouse and rat origins.

Type: Primary
Antigen: FIAF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR