Tisk…

Anti-SGLT-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3690-100EA 8740 CZK
BIOV3690-100
Anti-SGLT-2 Rabbit Polyclonal Antibody
Protilátky
Sodium glucose co transporters (SGLTs) are located in the brush-border membrane, but transported out of the cell across the basolateral membranes by a facilitated sμgar transporter (GLUT;At least three members of SGLTs (SGLT 1-3) have been cloned and characterized. SGLTs transport a methyl-D-glucoside (a-MDG), a non-metabolized model substrate, in Na-dependent manner. SGLT-1 does not discriminate a-MDG, glucose, and galactose. SGLT2/3 do not transport D-galactose efficiently.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SGLT-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR