Tisk…

Anti-HAT-2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3692-100EA 6910 CZK
BIOV3692-100
Anti-HAT-2 Rabbit Polyclonal Antibody
Protilátky
Histone acetyltransferases (HATs) have been implicated in a number of cellular functions including gene regulation, DNA synthesis, and repair. Histone acetyltransferases and deacetylases are, respectively, the enzymes devoted to the addition and removal of acetyl groups from lysine residues on the histone N-terminal tails. The enzymes exert fundamental roles in developmental processes and their deregulation has been linked to the progression of diverse human disorders, including cancer.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: HAT-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR