Tisk…

Anti-NCS-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3693-100EA 6910 CZK
BIOV3693-100
Anti-NCS-1 Rabbit Polyclonal Antibody
Protilátky
The neuronal calcium sensor (NCS) protein binds to calcium throμgh EF-hand motifs, are expressed in neurons. They are implicated in the regulation of neurotransmitters released at the neuromuscular junctions. NCS-1 is widely distributed in the brain, spinal cord and dorsal root ganglia. It has been shown to regulate neurosecreation in a calcium-dependent manner, potentiates nitric oxide synthase activity. NCS-1 has been reserved to indicate the protein observed in humans and mammals.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: NCS-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR