Tisk…

Anti-Bcl-B Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3695-100EA 7780 CZK
BIOV3695-100
Anti-Bcl-B Rabbit Polyclonal Antibody
Bcl-B (also called Bcl2-L-10) is a novel anti-apoptotic member of the Bcl-2 family. Bcl-B is closest in amino acid sequence homology to Boo protein. It contains four Bcl-2 homologs. Bcl-B mRNA is widely expressed in adult human tissues. The Bcl-B protein binds to Bcl-2, Bcl-X(L), and Bax but not Bak. Bcl-B suppresses apoptosis induced by Bax, but not by Bak.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Bcl-B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR