Tisk…

Anti-XAF-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3696-100EA 6910 CZK
BIOV3696-100
Anti-XAF-1 Rabbit Polyclonal Antibody
Protilátky
XAF-1 binds to XIAP, an inhibitor of caspases-3, -7, -9 and triggers its relocation from the cytosol to the nucleus. Overexpression of XAF-1 results in the neutralization of XIAP’s ability to inhibit cell death. XAF-1is normally expressed in all adult and fetal tissues but was found to be present in very low levels in a variety of cancer cell lines. In contrast, XIAP levels have been shown to be high in a majority of cell lines.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: XAF-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR