Tisk…

Anti-Dicer Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3697-100EA 6910 CZK
BIOV3697-100
Anti-Dicer Rabbit Polyclonal Antibody
Protilátky
Dicer is an RNaseIII-like enzyme which cleaves double-stranded RNA into 21-23 nucleotide (nt) RNAs. These RNAs are referred to as siRNA (short interfering RNA;It is believed to be essential in siRNA-mediated gene suppression.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Dicer
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR