Tisk…

Anti-MST-2/Krs-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3698-100EA 6910 CZK
BIOV3698-100
Anti-MST-2/Krs-1 Rabbit Polyclonal Antibody
Protilátky
Krs-1 protein exhibits stress-induced activity in response to staurosporine and okadaic acid. When the kinase-inactive mutant form of MST/Krs protein was overexpressed in cytotrienin A-sensitive HL-60 cells, the cytotrienin A-induced apoptosis was partially inhibited.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MST-2/Krs-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR