Tisk…

Anti-c-Jun Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3702-100EA 7470 CZK
BIOV3702-100
Anti-c-Jun Rabbit Polyclonal Antibody
Protilátky
Extracellular stimuli such as growth factors and UV irradiation stimulate phosphorylation of c-Jun at Ser63/73 and activate c-Jun dependent transcription. Mutation at Ser63/73 renders c-Jun nonresponsive to mitogenic and stress induced signaling pathways. JNK (c-Jun N-terminal Kinase) binds to the N-terminal region of c-Jun and phosphorylates c-Jun at Ser63/73. The activity of JNK is stimulated by the same signals that activate c-Jun.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml; Immunohistochemistry: 20-40 µg/ml

Type: Primary
Antigen: c-Jun
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR