Tisk…

Anti-BIK Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3703-100EA 6910 CZK
BIOV3703-100
Anti-BIK Rabbit Polyclonal Antibody
Protilátky
BIK is a member of Pro-apoptotic Bcl-2 family proteins. Althoμgh Bik lacks Bcl-2 homologus region, BH1 and BH2, BIK interacts with Bcl-2 and Bcl-xL. BIK has been shown to induce apoptosis by activating caspase-9.

Recommended Dilutions: Western Blot: 2-6 µg/ml

Type: Primary
Antigen: BIK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR