Tisk…

Anti-TUCAN/CARD8 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3705-100EA 6910 CZK
BIOV3705-100
Anti-TUCAN/CARD8 Rabbit Polyclonal Antibody
Protilátky
TUCAN (Tumor Up-regulated Card-containing Antagonist of Caspase-Nine) is a CARD (Caspase-Associated Recruitment Domains) containing protein that binds and suppresses activation of pro-Caspase-9 and found to be overexpressed in some types of cancers. TUCAN binds specifically to pro-caspase-9 and interferes with its interaction with Apaf-1, thereby suppressing apoptosis signaling in the mitochondrial/cytochrome c pathway.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TUCAN/CARD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR