Tisk…

Anti-Tubulin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3708-100EA 8180 CZK
BIOV3708-100
Anti-Tubulin Rabbit Polyclonal Antibody
Protilátky
Microtubules are one type of cytoskeleton. Globular tubulin subunits are the building blocks of a microtubule and the α and β-tubulin heterodimer forms the tubulin subunits, which is present in all eukaryotic cells. Microtubules mediate many cell movements.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Tubulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR