Tisk…

Anti-FOXP3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3709-100EA 6910 CZK
BIOV3709-100
Anti-FOXP3 Rabbit Polyclonal Antibody
Protilátky
FOXP3 a member of the forkhead/winge-helix family of transcriptional regulators and is highly conserved in humans. It has been proposed that FOXP3 may be a master regulatory gene and marker for Treg cells. Recent reports also sμggest expression of FOXP3 on subpopulation of rat CD8+ cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FOXP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR