Tisk…

Anti-Cathepsin V Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3712-100EA 6910 CZK
BIOV3712-100
Anti-Cathepsin V Rabbit Polyclonal Antibody
Protilátky
Cathepsin V, also known as Cathepsin L2 and Cathepsin U, is a cysteine protease of the papain family. It is expressed as a proenzyme in lysosomes, which can be autocatalytically converted to the mature form at pH 4.0. Cathepsin V is specifically expressed in the thymus, testis, and corneal epithelium.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: Cathepsin V
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR