Tisk…

Anti-DMAP1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3715-100EA 6910 CZK
BIOV3715-100
Anti-DMAP1 Rabbit Polyclonal Antibody
Protilátky
DNA methylation can help regulate transcriptional silencing via repressive transcription complexes, which include methyl-CpG-binding domain protein and Histone deacetylases (HDACs) DNMT1. This core enzyme for mammalian DNA methylation can also establish a repressive transcription complex consisting of DNMT1, HDAC2 and DMAP1. DMAP1 can interact directly with the transcriptional corepressor TSG101. The DNMT1-DMAP1 exists throμghout the S-phase; HDAC2 joind DNMT1 and DMAP1 only during Late S phase. This provides a regulated means to deacetylate heterochromatin following replication.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DMAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR