Tisk…

Anti-RBP4 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3716-100EA 8430 CZK
BIOV3716-100
Anti-RBP4 Rabbit Polyclonal Antibody
RBP4 (Retinol Binding Protein 4;Retinol is transported in blood bound to the carrier protein RBP. Under physiological conditions, RBP binds transthyretin (TTR) to prevent glomerular filtration of low molecular weight RBP in the kidneys. The RBP/TTR molar ration provides an indirect way to indicate marginal vitamin A deficiency.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: RBP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR