Tisk…

Anti-VASP Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3718-100EA 6910 CZK
BIOV3718-100
Anti-VASP Rabbit Polyclonal Antibody
Protilátky
Vasodilator-stimulated phosphoprotein (VASP) belongs to the Ena/VASP family of adaptor proteins linking the cytoskeletal system to the signal transduction pathways. VASP functions in cytoskeletal organization, fibroblast migration, platelet activation and axon guidance. Three phosphorylation sites, Ser157, Ser239 and Thr278, have been identified. Evidence sμggests that VASP phosphorylation reduces its association with actin and has a negative effect on actin polymerization.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: VASP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR