Tisk…

Anti-Kallikrein 12 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3720-100EA 6910 CZK
BIOV3720-100
Anti-Kallikrein 12 Rabbit Polyclonal Antibody
Protilátky
The human KLK gene family encodes about 15 serine proteases and has the potential to serve as cancer biomarkers. The Kallikrein 12 (KLK 12) is one the KLK members located on chromosome 19. Different splice variants of the KLK12 produces three transcript variants encoding different isoforms that can be found on amino acid 254, 248 and 110.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Kallikrein 12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR