Tisk…

Anti-CRSP1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3735-100EA 10970 CZK
BIOV3735-100
Anti-CRSP1 Rabbit Polyclonal Antibody
CSP1/TRAP220 is a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.

Recommended Dilutions: Western Blot: 1:500-1:2500 µg/ml

Type: Primary
Antigen: CRSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR