Tisk…

Anti-Visfatin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5909-100EA 6910 CZK
BIOV5909-100
Anti-Visfatin Rabbit Polyclonal Antibody
Protilátky
Visfatin is a cytokine highly expressed in visceral fat and blood. This protein has also been reported to be a cytokine (PBEF) that promotes B cell maturation and inhibits neutrophil apoptosis. Visfatin is thoμght to have insulin-like activities and is able to bind insulin receptor and thus, may lead to lowering blood glucose levels.

Recommended Dilutions: Western blot: 0.5-4 µg/ml

Type: Primary
Antigen: Visfatin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR