Tisk…

Anti-Ghrelin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5990-100EA 6910 CZK
BIOV5990-100
Anti-Ghrelin Rabbit Polyclonal Antibody
Protilátky
Ghrelin, a growth hormone (GH)-releasing acylated peptide, was identified as the endogenous ligand for the growth hormone (GH) secretagogue receptor 1a. It stimulates the release of GH from the anterior pituitary throμgh the GH secretagogue receptor (GHS-R;It may also play a role in regulating the digestive system and energy balance.

Recommended Dilutions:

Type: Primary
Antigen: Ghrelin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR