Tisk…

Anti-mCherry Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV5993-100EA 9590 CZK
BIOV5993-100
Anti-mCherry Rabbit Polyclonal Antibody
Protilátky
mCherry is the second generation monomeric red fluorescent protein that have improved brightness and photostability. The recombinant mCherry is expressed and purified from transformed E. coli using a method that ensures high purity and maximal fluorescence intensity. mCherry protein is a 28.8 kDa monomer with 256 amino acid.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: mCherry
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR