Tisk…

Anti-Histone H3 (phospho-Ser28) Mouse Monoclonal Antibody [clone: 117C826]

Dodavatel: BioVision
BIOV6119-50EA 6910 CZK
BIOV6119-50
Anti-Histone H3 (phospho-Ser28) Mouse Monoclonal Antibody [clone: 117C826]
Protilátky
The phosphorylation of histone H3 plays an important role in gene expression, chromatin remodeling, chromosome condensation, and cell division. Histone H3 phosphorylation may initiate at different phases of the cell division in different organisms, but metaphase chromosomes are always found to be heavily phosphorylated. H3 phosphorylation at serine 28 is coupled with mitotic chromosome condensation in diverse mammalian cell lines.

Recommended Dilutions: Western Blot: 2 µg/ml

Type: Primary
Antigen: Histone H3 (phospho-Ser28)
Clonality: Monoclonal
Clone: 117C826
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR