Tisk…

Anti-Acetyl Lysine Rabbit Polyclonal Antibody (Biotin)

Dodavatel: BioVision
BIOV6125-50EA 7780 CZK
BIOV6125-50
Anti-Acetyl Lysine Rabbit Polyclonal Antibody (Biotin)
Post-translational modifications of proteins play critical roles in the regulation and function of many known biological processes. Proteins can be post-translationally modified in many different ways, and a common post-transcriptional modification of lysine involves acetylation. The conserved amino-terminal domains of the four core histones (H2A, H2B, H3, and H4) contain lysines that are acetylated by histone acetyltransferases (HATs) and deacetylated by histone deacetylases (HDACs;Protein post-translational reversible lysine Nε-acetylation and deacetylation have been recognized as an emerging intracellular signaling mechanism that plays critical roles in regulating gene transcription, cell-cycle progression, apoptosis, DNA repair, and cytoskeletal organization. The regulation of protein acetylation status is impaired in the pathologies of cancer and polyglutamine diseases, and HDACs have become promising targets for anti-cancer drμgs currently in development.

Recommended Dilutions: Western blot, IP, ELISA and IF.

Type: Primary
Antigen: Acetyl Lysine
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Multiple Species
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR