Tisk…

Anti-NIF1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3737R-100EA 6910 CZK
BIOV3737R-100
Anti-NIF1 Rabbit Polyclonal Antibody
Protilátky
NRC-Interacting Factor 1 (NIF1) interacts with the nuclear receptor transcriptional coactivator NRC and enhances ligand-dependent transcriptional activation by nuclear hormone receptors as well as c-Fos and c-Jun. NIF1 may function to influence chromatin structure.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: NIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR