Tisk…

Anti-FoxL2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3742-100EA 6910 CZK
BIOV3742-100
Anti-FoxL2 Rabbit Polyclonal Antibody
Protilátky
FOXL2 is a nuclear protein specially expressed in eyelids and in fetal and adult ovarian follicular cells. Defects in FOXL2 are cause of blepharophimosis, ptosis and epicanthus inversus syndrome (BPES;It is an autosomal dominant disorder characterized by eyelid dysplasia, small palpebral fissures, drooping eyelids and a skin fold. It is also associated with female infertility.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FoxL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR