Tisk…

Anti-SMC2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3754-100EA 6910 CZK
BIOV3754-100
Anti-SMC2 Rabbit Polyclonal Antibody
Protilátky
Structural maintenance of chromosomes (SMC) proteins are part of the cohesin and condensin multisubunit complexes that play a critical role in chromosome organization, segregation, gene regulation, and DNA repair. The core condensin complex includes a heterodimer of SMC2 and SMC4 that functions to create positively supercoiled loops for chromatin condensation and metaphase chromatid shaping. A function for condensin in the DNA damage checkpoint response and gene repression has also been suggested.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: SMC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR