Tisk…

Anti-XLF Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3757-100EA 6910 CZK
BIOV3757-100
Anti-XLF Rabbit Polyclonal Antibody
Protilátky
XLF (also called Cernunnos) interacts with the XRCC4-DNA ligase IV complex to promote DNA non-homologous end-joining. It directly interacts with the XRCC4-Ligase IV complex and siRNA-mediated downregulation of XLF in human cell lines leads to radio-sensitivity and impaired DNA non-homologous end-joining. XLF is homologous to the yeast non-homologous end-joining factor Nej1.

Recommended Dilutions: Western Blot: 1:500-1:2500; Immunoprecipitation

Type: Primary
Antigen: XLF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR