Tisk…

Anti-MCAK Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3760-100EA 7780 CZK
BIOV3760-100
Anti-MCAK Rabbit Polyclonal Antibody
Kinesin activity has been linked to various cellular functions such as vesicle transport, mitotic spindle formation, chromosome segregation, chromosome congression, and cytokinesis. Structurally, all kinesins contain a motor domain with microtubule and nucleotide binding sites that utilize ATP to target cargo along microtubule filaments. MCAK (Mitotic Centromere-Associated Kinesin) is the founding member of kinesin-13 proteins. There are 3 independent genes coding for kinesin-13 proteins kif2a, kif2b, and kif2c (MCAK;Instead of functioning as a translocator of microtubules MCAK is a depolymerizer that is primarily responsible for releasing improper microtubule-kinetochore attachments during cell division.

Recommended Dilutions: Western Blot: 1:500-1:2000

Type: Primary
Antigen: MCAK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR