Tisk…

Anti-KAP-1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3764-100EA 6910 CZK
BIOV3764-100
Anti-KAP-1 Rabbit Polyclonal Antibody
Protilátky
KAP-1 (KRAB-Associated Protein 1) is a member of the tripartite motif RBCC/Trim domain family that contains a RING finger domain, B boxes, and an alpha helical coiled coil region. KAP-1 functions as a corepressor by interacting with the KRAB domain of KRAB zinc-finger transcriptional repressors. KAP-1 also appears to function as a scaffold for chromatin-remodeling complexes involved in transcriptional repression.

Recommended Dilutions: Western Blot: 1:500-1:2500

Type: Primary
Antigen: KAP-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR