Tisk…

Anti-SRC3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3773-100EA 6910 CZK
BIOV3773-100
Anti-SRC3 Rabbit Polyclonal Antibody
Protilátky
Steroid receptor coactivator 3 (SRC3) belongs to the SRC family of proteins that interacts with nuclear receptors to facilitate transcription. The 3 homologous members of this family are SRC-1 (NCoA-1), SRC-2 (GRIP1, TIF2, or NcoA-2), and SRC-3 (p/CIP, RAC3, ACTR, AIB1, or TRAM-1;SRC3 is an oncogene that is reported to be amplified and/or overexpressed in breast and gastric cancer and stimulates AKT signaling and cell growth and tumorigenesis.

Recommended Dilutions: Western Blot: 1:500-1:2500

Type: Primary
Antigen: SRC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR