Tisk…

Anti-USP2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6141-100EA 9450 CZK
BIOV6141-100
Anti-USP2 Rabbit Polyclonal Antibody
Protilátky
Ubiquitinating enzymes (UBEs) catalyze protein ubiquitination, a reversible process countered by deubiquitinating enzymes (DUB) action. There are five DUB subfamilies including the USP, UCH, OTU, MJD, and JAMM enzymes. Ubiquitin­specific­processing protease 2 (USP2) belongs to the USP (UBP/UCH type 2) subfamily and is characterized by its C19 peptidase activity, which is involved in ubiquitin recycling and in the disassembly of various forms of polymeric ubiquitin and ubiquitin like protein complexes. There is mounting evidence that USP2 functions as an oncoprotein through multiple mechanisms. In human prostate cancer, USP2 is highly overexpressed and drives tumor growth by binding to and stabilizing fatty acid synthase through deubiquitination. It has also been demonstrated that USP2 can bind and deubiquitinate both Mdm2 and cyclin D1, which leads to their stabilization and enhanced cell proliferation.

Recommended Dilutions: Western Blot: 1:200-1:500 dilution, IHC: 1:50 - 1:100 dilution. Immunofluorescence: 1:20 - 1:50 dilution

Type: Primary
Antigen: USP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR