Tisk…

Anti-UHRF1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6144-100EA 9450 CZK
BIOV6144-100
Anti-UHRF1 Rabbit Polyclonal Antibody
Protilátky
UHRF1, also named as ICBP90, NP95 and RNF106, is a putative E3 ubiquitin protein ligase. It may participate in methylation dependent transcriptional regulation. UHRF1 binds to inverted 5'­CCAAT­3' box 2 in the TOP2A promoter, and activates TOP2A expression. It is important for G1/S transition and may be involved in DNA repair and chromosomal stability. UHRF1's ability to repress its direct target gene expression is dependent on PHD (UHRF1) binding to unmodified H3R2.

Recommended Dilutions: Western Blot: 1:500-1:2000, IHC: 1:50-1:200, Immunofluorescence: 1:20-1:50, IP: 1:20-1:50, CHIP: 1:20-1:50

Type: Primary
Antigen: UHRF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR