Tisk…

Anti-UHRF2 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6145-100EA 9450 CZK
BIOV6145-100
Anti-UHRF2 Rabbit Polyclonal Antibody
Protilátky
This gene encodes a nuclear protein which is involved in cell cycle regulation. The encoded protein is a ubiquitin ligase capable of ubiquinating PCNP (PEST­containing nuclear protein), and together they may play a role in tumorigenesis. The encoded protein contains an NIRF_N domain, a PHD finger, a set­ and ring associated (SRA) domain, and a RING finger domain and several of these domains have been shown to be essential for the regulation of cell proliferation. This protein may also have a role in intranuclear degradation of polyglutamine aggregates. Alternative splicing results in multiple transcript variants, some of which are non­protein coding. [Provided by RefSeq, Feb 2012].

Recommended Dilutions: Western Blot: 1:500-1:2000, IHC: 1:50-1:200, Immunofluorescence: 1:20-1:50.

Type: Primary
Antigen: UHRF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR