Tisk…

Anti-CBX3 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6147-100EA 9450 CZK
BIOV6147-100
Anti-CBX3 Rabbit Polyclonal Antibody
Protilátky
Heterochromatin protein 1 (HP1) is a family of heterochromatic adaptor molecules involved in both gene silencing and higher order chromatin structure. All three HP1 family members (α, β, and γ) are primarily associated with centromeric heterochromatin; However, HP1β and γ also localize to euchromatic sites in the genome. HP1 proteins are approximately 25 kDa in size and contain a conserved amino terminal chromodomain, followed by a variable hinge region and a conserved carboxy­terminal chromoshadow domain. The chromodomain facilitates binding to histone H3 tri­methylated at Lys9, a histone "mark" closely associated with centromeric heterochromatin. The variable hinge region binds both RNA and DNA in a sequence independent manner. The chromoshadow domain mediates the dimerization of HP1 proteins, in addition to binding multiple proteins  implicated in gene silencing and heterochromatin  formation, including the SUV39H histone methyltransferase, the DNMT1 and DNMT3a DNA  methyltransferases, and the p150 subunit of chromatin­assembly factor­1 (CAF1;In addition to contributing to heterochromatin formation and propagation, HP1 and SUV39H are also found complexed with retinoblastoma (Rb) and E2F6 proteins, both of which function to repress euchromatic gene transcription in quiescent cells. HP1 proteins are subject to multiple types of post­translational modifications, including phosphorylation, acetylation, methylation, ubiquitination, and sumoylation, suggesting multiple means of regulation.

Recommended Dilutions: Western Blot: 1:500-1:2000, IHC: 1:50-1:100, Immunofluorescence: 1:20-1:50

Type: Primary
Antigen: CBX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR