Tisk…

Anti-NCOA1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV6153-100EA 9450 CZK
BIOV6153-100
Anti-NCOA1 Rabbit Polyclonal Antibody
Protilátky
There are three members of the steroid receptor co­activator (SRC) family of proteins: SRC­1 (NCoA­1), SRC­2 (TIF2/GRIP1/NCoA­2), and SRC­3 (ACTR /TRAM­1/AIB1;All SRC family members share significant structural homology and function to stimulate transcription mediated by nuclear hormone receptors and other transcriptional activators such as Stat3, NF­κB, E2F1, and p53. Two SRC proteins, SRC­1 and SRC­3, function as histone acetyltransferases. In addition, all three family members can recruit other histone acetyltransferases (CBP/p300, PCAF) and histone methyltransferases (PRMT1, CARM1) to target promoters and cooperate to enhance expression of many genes. The SRC proteins play important roles in multiple physiological processes including cell proliferation, cell survival, somatic cell growth, mammary gland development, female reproductive function, and vasoprotection. SRC­1 and SRC­3 are conduits for kinase mediated growth factor signaling to the estrogen receptor and other transcriptional activators. Seven SRC­1 phosphorylation sites and six SRC­3 phosphorylation sites have been identified, which are induced by steroids, cytokines, and growth factors and involve multiple kinase signaling pathways. Research has shown that all three SRC family members are associated with increased activity of nuclear receptors in breast, prostate, and ovarian carcinomas. According to the literature, SRC­3 is frequently amplified or overexpressed in a number of cancers, and SRC­1/PAX3 and SRC­2/MYST3 translocations are found associated with rhabdomyosarcoma and acute myeloid leukemia, respectively.

Recommended Dilutions: Western Blot: 1:500-1:2000, IHC: 1:50-1:200.

Type: Primary
Antigen: NCOA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR