Tisk…

Anti-TAOK1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3776R-100EA 6910 CZK
BIOV3776R-100
Anti-TAOK1 Rabbit Polyclonal Antibody
Protilátky
Serine/threonine-protein kinase TAO1 is an enzyme that in humans is encoded by the TAOK1 gene.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TAOK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR