Tisk…

Anti-RCH1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3779-100EA 9730 CZK
BIOV3779-100
Anti-RCH1 Rabbit Polyclonal Antibody
Protilátky
The import of proteins from the cytoplasm to the nucleus involves docking the protein to receptors associated with the nuclear pore complex followed by translocation through the nuclear pore. RCH1 is an adaptor protein that recognizes the nuclear localization signal (NLS) on cargo and binds the karyopherin importin-beta receptor for nuclear import which is regulated by RanGTP.

Recommended Dilutions: Western Blot: 1:500-1:2000; Immunoprecipitation

Type: Primary
Antigen: RCH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR