Tisk…

Anti-SRA Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3780-100EA 9730 CZK
BIOV3780-100
Anti-SRA Rabbit Polyclonal Antibody
Protilátky
Steroid receptor RNA activator (SRA) selectively mediates transactivation of steroid hormone receptors. Specifically, SRA exists as both an RNA transcript that forms a complex with steroid receptor coactivator-1 and as a stably expressed protein. There are six RNA motifs in SRA that are important for coactivation. SRA is ubiquitously expressed in normal tissues with higher levels of expression in liver and skeletal muscle. SRA is expressed at a low level in brain. SRA is expressed at higher levels in breast tumor than in normal tissue. Overexpression of SRA stimulates ERalpha transcriptional activity. In cells transfected with antisense oligodeoxynucleotides to SRA, ERα expression is reduced in a dose-dependent fashion. SMRT/HDAC1 associated repressor protein (SHARP) binds to SRA and inhibits SRA-potentiated steroid receptor transciption.

Recommended Dilutions: Western Blot: 1:500-1:2500

Type: Primary
Antigen: SRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR