Tisk…

Anti-PIM1 Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3787R-100EA 6910 CZK
BIOV3787R-100
Anti-PIM1 Rabbit Polyclonal Antibody
Protilátky
The proto-oncogene Pim1 belongs to a family of serine/threonine protein kinases that are highly conserved throμgh evolution in multicellular organisms. Pim1 is involved in the control of cytokine-mediated cell proliferation, differentiation and survival of lymphoid and myeloid cells as well as others. Expression of Pim1 can be stimulated by a variety of growth factors and is regulated at four different levels: transcriptional, post-transcriptional, translational and post-translational. Recently, Pim1 has been shown to enhance the activities of p100, c-Myb and Cdc 25a and in part this might explain reported effects of Pim1 on mitogenesis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR