Tisk…

Anti-Prohibitin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3805-100EA 6910 CZK
BIOV3805-100
Anti-Prohibitin Rabbit Polyclonal Antibody
Protilátky
Prohibitin is an evolutionarily conserved protein that has anti-proliferative activity. Prohibitin is a 30 kDa, post-synthetically modified protein found in the inner membrane of mitochondria and on the plasma membrane of B cells. It mediates B cell mutation. Prohibitin Controls cell cycle in both transcriptional and post-translational mechanisms. Prohibitin is a potential tumor suppressor. Prohibitin gene mutations are correlated with the breast cancer development and/or progression in over 80%0f the cell lines analyzed.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 10-20 µg/ml; Iimmunofluorescence: 5-10 µg/ml

Type: Primary
Antigen: Prohibitin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR