Tisk…

Anti-Catalase Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3806-100EA 6910 CZK
BIOV3806-100
Anti-Catalase Rabbit Polyclonal Antibody
Protilátky
Catalase is known marker for peroxisomes. It is the most abundant protein in the peroxisomes. It is present in all aerobically respiring organisms. It protects cells form the toxicity of hydrogen peroxide.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Catalase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR