Tisk…

Anti-Calnexin Rabbit Polyclonal Antibody

Dodavatel: BioVision
BIOV3811-100EA 6910 CZK
BIOV3811-100
Anti-Calnexin Rabbit Polyclonal Antibody
Protilátky
Calnexin (CNX) is a 90kDa integral protein of the endoplasmic reticulum (ER;Calnexin belongs among chaperones, which are characterized by their main function of assisting protein folding and quality control, ensuring that only properly folded and assembled proteins proceed further along the secretary pathway. The function of Calnexin is to retain unfolded or unassembled N-linked glycoproteins in the endoplasmic reticulum.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Calnexin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR